Assay Details
| Gene Symbol: qki | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100198.1 | NP_001093668.1 | 5 - 6 | 1118 | 95 | |
| XM_012962554.2 | XP_012818008.1 | 6 - 7 | 1353 | 95 | ||
| XM_012962555.2 | XP_012818009.1 | 6 - 7 | 1354 | 95 | ||
| XM_012962556.2 | XP_012818010.1 | 6 - 7 | 1355 | 95 | ||
| XM_012962557.2 | XP_012818011.1 | 5 - 6 | 1305 | 95 | ||
| XM_012962558.2 | XP_012818012.1 | 5 - 6 | 1306 | 95 | ||
| XM_012962559.2 | XP_012818013.1 | 6 - 7 | 1355 | 95 | ||
| XM_012962560.2 | XP_012818014.1 | 5 - 6 | 1306 | 95 | ||
| XM_012962561.2 | XP_012818015.1 | 5 - 6 | 1306 | 95 | ||
| XM_012962562.2 | XP_012818016.1 | 5 - 6 | 1308 | 95 | ||
| XM_018093738.1 | XP_017949227.1 | 7 - 8 | 973 | 95 | ||
| XM_018093739.1 | XP_017949228.1 | 6 - 7 | 1354 | 95 | ||
| GenBank mRNA | BC135440.1 | - | 5 - 6 | 1118 | 95 | |
More Information
Set Membership: |
|

qki
Genomic Map