Assay Details
| Gene Symbol: nr4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100208.1 | NP_001093678.1 | 4 - 5 | 1144 | 61 | |
| XM_012969938.2 | XP_012825392.1 | 4 - 5 | 1115 | 61 | ||
| GenBank mRNA | BC136009.1 | - | 4 - 5 | 1144 | 61 | |
More Information
Set Membership: |
|

nr4a2
Genomic Map