Assay Details
| Gene Symbol: dll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100219.1 | NP_001093689.1 | 4 - 5 | 1197 | 91 | |
| GenBank mRNA | BC135592.1 | - | 4 - 5 | 1197 | 91 | |
More Information
Set Membership: |
|

dll1
Genomic Map