Assay Details
| Gene Symbol: hand2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100225.1 | NP_001093695.1 | 1 - 2 | 1112 | 90 | |
| XM_012960031.2 | XP_012815485.1 | 2 - 3 | 885 | 90 | ||
| GenBank mRNA | BC135783.1 | - | 1 - 2 | 1112 | 90 | |
More Information
Set Membership: |
|

hand2
Genomic Map