Assay Details
| Gene Symbol: fscn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100254.1 | NP_001093724.1 | 2 - 3 | 1057 | 70 | |
| GenBank mRNA | BC135752.1 | - | 2 - 3 | 1057 | 70 | |
More Information
Set Membership: |
|

fscn2
Genomic Map