Assay Details
| Gene Symbol: egr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100255.1 | NP_001093725.1 | 1 | 382 | 101 | |
| GenBank mRNA | BC136205.1 | - | 1 | 382 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

egr2| Gene Symbol: egr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100255.1 | NP_001093725.1 | 1 | 382 | 101 | |
| GenBank mRNA | BC136205.1 | - | 1 | 382 | 101 | |
Set Membership: |
|