Assay Details
| Gene Symbol: nr1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100258.1 | NP_001093728.1 | 2 - 3 | 625 | 64 | |
| GenBank mRNA | BC135445.1 | - | 2 - 3 | 625 | 64 | |
More Information
Set Membership: |
|

nr1d2
Genomic Map