Assay Details
| Gene Symbol: onecut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100260.1 | NP_001093730.1 | 1 - 2 | 1302 | 80 | |
| GenBank mRNA | BC135775.2 | - | 1 - 2 | 1302 | 80 | |
More Information
Set Membership: |
|

onecut1
Genomic Map