Assay Details
| Gene Symbol: tnfrsf11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012965431.2 | XP_012820885.1 | 7 - 8 | 1294 | 68 | |
| GenBank mRNA | BC136133.1 | - | 6 - 7 | 1577 | 68 | |
More Information
Set Membership: |
|

tnfrsf11a
Genomic Map