Assay Details
| Gene Symbol: ankrd46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100266.1 | NP_001093736.1 | 3 - 4 | 490 | 76 | |
| XM_012964168.2 | XP_012819622.1 | 3 - 4 | 511 | 76 | ||
| GenBank mRNA | BC136012.1 | - | 3 - 4 | 490 | 76 | |
More Information
Set Membership: |
|

ankrd46
Genomic Map