Assay Details
| Gene Symbol: ppp2r3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002939919.4 | XP_002939965.2 | 3 - 4 | 802 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ppp2r3a| Gene Symbol: ppp2r3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002939919.4 | XP_002939965.2 | 3 - 4 | 802 | 65 | |
Set Membership: |
|