Assay Details
Gene Symbol: rab3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100276.1 | NP_001093746.1 | 2 - 3 | 361 | 79 | |
XM_018092876.1 | XP_017948365.1 | 2 - 3 | 304 | 79 | ||
GenBank mRNA | BC135348.1 | - | 2 - 3 | 361 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
