Assay Details
| Gene Symbol: rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100276.1 | NP_001093746.1 | 4 - 5 | 607 | 93 | |
| XM_018092876.1 | XP_017948365.1 | 4 - 5 | 550 | 93 | ||
| GenBank mRNA | BC135348.1 | - | 4 - 5 | 607 | 93 | |
More Information
Set Membership: |
|

rab3b
Genomic Map