Assay Details
| Gene Symbol: hdhd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100283.1 | NP_001093753.1 | 1188 | 83 | ||
| GenBank mRNA | BC136143.1 | - | 1188 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hdhd5| Gene Symbol: hdhd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100283.1 | NP_001093753.1 | 1188 | 83 | ||
| GenBank mRNA | BC136143.1 | - | 1188 | 83 | ||
Set Membership: |
|