Assay Details
| Gene Symbol: siah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101811.1 | NP_001095281.1 | 1 - 2 | 543 | 61 | |
| GenBank mRNA | BC136087.1 | - | 1 - 2 | 543 | 61 | |
More Information
Set Membership: |
|

siah2
Genomic Map