Assay Details
| Gene Symbol: ube2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101813.1 | NP_001095283.1 | 1 - 2 | 237 | 62 | |
| GenBank mRNA | BC127550.1 | - | 1 - 2 | 237 | 62 | |
| CT025222.2 | - | 1 - 2 | 154 | 62 | ||
More Information
Set Membership: |
|

ube2c
Genomic Map