Assay Details
| Gene Symbol: rab11fip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012959457.2 | XP_012814911.1 | 5 - 6 | 3196 | 68 | |
| GenBank mRNA | BC135580.1 | - | 4 - 5 | 1691 | 68 | |
| CR942770.1 | - | 5 - 6 | 2961 | 68 | ||
More Information
Set Membership: |
|

rab11fip1
Genomic Map