Assay Details
| Gene Symbol: dnajc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102684.1 | NP_001096154.1 | 1292 | 61 | ||
| GenBank mRNA | BC129014.1 | - | 1292 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dnajc27| Gene Symbol: dnajc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102684.1 | NP_001096154.1 | 1292 | 61 | ||
| GenBank mRNA | BC129014.1 | - | 1292 | 61 | ||
Set Membership: |
|