Assay Details
| Gene Symbol: zdhhc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102692.1 | NP_001096162.1 | 3 - 4 | 551 | 76 | |
| XM_012967852.2 | XP_012823306.1 | 3 - 4 | 561 | 76 | ||
| GenBank mRNA | BC136005.1 | - | 3 - 4 | 551 | 76 | |
More Information
Set Membership: |
|

zdhhc9
Genomic Map