Assay Details
| Gene Symbol: gabrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102694.1 | NP_001096164.1 | 5 - 6 | 971 | 78 | |
| GenBank mRNA | BC136049.1 | - | 5 - 6 | 971 | 78 | |
More Information
Set Membership: |
|

gabrb3
Genomic Map