Assay Details
| Gene Symbol: rel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102707.1 | NP_001096177.1 | 9 - 10 | 1350 | 70 | |
| GenBank mRNA | BC136176.1 | - | 9 - 10 | 1350 | 70 | |
More Information
Set Membership: |
|

rel
Genomic Map