Assay Details
| Gene Symbol: slc25a29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102721.1 | NP_001096191.1 | 2 - 3 | 320 | 74 | |
| GenBank mRNA | BC136177.1 | - | 2 - 3 | 320 | 74 | |
More Information
Set Membership: |
|

slc25a29
Genomic Map