Assay Details
| Gene Symbol: csgalnact2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102722.1 | NP_001096192.1 | 5 - 6 | 1521 | 62 | |
| XM_012965870.2 | XP_012821324.1 | 5 - 6 | 1592 | 62 | ||
| XM_012965871.2 | XP_012821325.1 | 7 - 8 | 1747 | 62 | ||
| XM_012965872.2 | XP_012821326.1 | 6 - 7 | 1693 | 62 | ||
| XM_012965874.2 | XP_012821328.1 | 6 - 7 | 1646 | 62 | ||
| XM_018095219.1 | XP_017950708.1 | 8 - 9 | 1861 | 62 | ||
| XM_018095220.1 | XP_017950709.1 | 6 - 7 | 1736 | 62 | ||
| GenBank mRNA | BC135663.1 | - | 5 - 6 | 1521 | 62 | |
More Information
Set Membership: |
|

csgalnact2
Genomic Map