Assay Details
| Gene Symbol: spock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102726.1 | NP_001096196.1 | 7 - 8 | 698 | 66 | |
| XM_012967860.2 | XP_012823314.1 | 7 - 8 | 762 | 66 | ||
| XM_012967861.2 | XP_012823315.1 | 6 - 7 | 752 | 66 | ||
| GenBank mRNA | BC135841.1 | - | 7 - 8 | 698 | 66 | |
More Information
Set Membership: |
|

spock3
Genomic Map