Assay Details
Gene Symbol: acbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102728.1 | NP_001096198.1 | 1 - 2 | 252 | 58 | |
XM_012962605.2 | XP_012818059.1 | 1 - 2 | 358 | 58 | ||
GenBank mRNA | BC135953.1 | - | 1 - 2 | 252 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
