Assay Details
| Gene Symbol: wasf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102747.1 | NP_001096217.1 | 6 - 7 | 932 | 64 | |
| XM_012962614.1 | XP_012818068.1 | 6 - 7 | 1288 | 64 | ||
| XM_018093759.1 | XP_017949248.1 | 4 - 5 | 620 | 64 | ||
| GenBank mRNA | BC091579.1 | - | 6 - 7 | 932 | 64 | |
More Information
Set Membership: |
|

wasf1
Genomic Map