Assay Details
| Gene Symbol: tpx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102763.1 | NP_001096233.1 | 327 | 70 | ||
| GenBank mRNA | BC135342.1 | - | 327 | 70 | ||
| CT027887.1 | - | 334 | 70 | |||
More Information
Set Membership: |
|

tpx2
Genomic Map