Assay Details
| Gene Symbol: myl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102780.1 | NP_001096250.1 | 3 - 4 | 432 | 60 | |
| XM_012971505.2 | XP_012826959.1 | 3 - 4 | 448 | 60 | ||
| GenBank mRNA | BC077710.1 | - | 3 - 4 | 425 | 60 | |
| BC135412.1 | - | 3 - 4 | 432 | 60 | ||
More Information
Set Membership: |
|

myl9
Genomic Map