Assay Details
| Gene Symbol: fabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102786.1 | NP_001096256.1 | 1 - 2 | 119 | 85 | |
| GenBank mRNA | BC135431.1 | - | 1 - 2 | 119 | 85 | |
More Information
Set Membership: |
|

fabp4
Genomic Map