Assay Details
| Gene Symbol: rem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102791.1 | NP_001096261.1 | 4 - 5 | 651 | 79 | |
| XM_012955294.2 | XP_012810748.1 | 4 - 5 | 2014 | 79 | ||
| GenBank mRNA | BC135453.1 | - | 4 - 5 | 651 | 79 | |
More Information
Set Membership: |
|

rem1
Genomic Map