Assay Details
| Gene Symbol: cdcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102795.1 | NP_001096265.1 | 4 - 5 | 1146 | 70 | |
| GenBank mRNA | BC135475.1 | - | 4 - 5 | 1146 | 70 | |
More Information
Set Membership: |
|

cdcp2
Genomic Map