Assay Details
| Gene Symbol: dpcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102805.1 | NP_001096275.1 | 4 - 5 | 596 | 80 | |
| GenBank mRNA | BC135507.1 | - | 4 - 5 | 596 | 80 | |
More Information
Set Membership: |
|

dpcd
Genomic Map