Assay Details
| Gene Symbol: serhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102810.2 | NP_001096280.2 | 8 - 9 | 675 | 59 | |
| GenBank mRNA | BC135528.1 | - | 8 - 9 | 675 | 59 | |
| CX499519.2 | - | 8 - 9 | 632 | 59 | ||
More Information
Set Membership: |
|

serhl2
Genomic Map