Assay Details
| Gene Symbol: guca1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102821.1 | NP_001096291.1 | 1 - 2 | 450 | 92 | |
| GenBank mRNA | BC135577.1 | - | 1 - 2 | 450 | 92 | |
More Information
Set Membership: |
|

guca1a
Genomic Map