Assay Details
| Gene Symbol: cbfa2t2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102826.1 | NP_001096296.1 | 2 - 3 | 516 | 68 | |
| GenBank mRNA | BC135600.1 | - | 2 - 3 | 516 | 68 | |
More Information
Set Membership: |
|

cbfa2t2
Genomic Map