Assay Details
| Gene Symbol: tsen15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102835.1 | NP_001096305.2 | 3 - 4 | 329 | 73 | |
| XM_012960462.2 | XP_012815916.1 | 3 - 4 | 576 | 73 | ||
| GenBank mRNA | BC135656.1 | - | 3 - 4 | 329 | 73 | |
More Information
Set Membership: |
|

tsen15
Genomic Map