Assay Details
| Gene Symbol: lgals9c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102848.1 | NP_001096318.1 | 8 - 9 | 800 | 88 | |
| XM_012956288.2 | XP_012811742.1 | 9 - 10 | 895 | 88 | ||
| XM_012956289.2 | XP_012811743.1 | 8 - 9 | 851 | 88 | ||
| XM_012956290.2 | XP_012811744.1 | 7 - 8 | 839 | 88 | ||
| GenBank mRNA | BC081298.1 | - | 8 - 9 | 806 | 88 | |
| BC135696.1 | - | 8 - 9 | 800 | 88 | ||
More Information
Set Membership: |
|

lgals9c
Genomic Map