Assay Details
| Gene Symbol: gck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102851.1 | NP_001096321.1 | 4 - 5 | 645 | 67 | |
| XM_012958422.2 | XP_012813876.1 | 4 - 5 | 938 | 67 | ||
| GenBank mRNA | BC135716.1 | - | 4 - 5 | 645 | 67 | |
More Information
Set Membership: |
|

gck
Genomic Map