Assay Details
| Gene Symbol: slc35a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102852.1 | NP_001096322.1 | 1 | 953 | 99 | |
| XM_012958423.1 | XP_012813877.1 | 2 | 1700 | 99 | ||
| GenBank mRNA | BC135720.1 | - | 1 | 953 | 99 | |
More Information
Set Membership: |
|

slc35a4
Genomic Map