Assay Details
| Gene Symbol: opn1lw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102861.1 | NP_001096331.1 | 1 - 2 | 220 | 87 | |
| GenBank mRNA | BC135755.1 | - | 1 - 2 | 220 | 87 | |
More Information
Set Membership: |
|

opn1lw
Genomic Map