Assay Details
| Gene Symbol: gpr176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102864.1 | NP_001096334.1 | 1 - 2 | 436 | 79 | |
| GenBank mRNA | BC135765.1 | - | 1 - 2 | 436 | 79 | |
More Information
Set Membership: |
|

gpr176
Genomic Map