Assay Details
| Gene Symbol: pagr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102877.1 | NP_001096347.1 | 2 - 3 | 624 | 93 | |
| XM_012954605.2 | XP_012810059.1 | 2 - 3 | 753 | 93 | ||
| GenBank mRNA | BC135816.1 | - | 2 - 3 | 624 | 93 | |
More Information
Set Membership: |
|

pagr1
Genomic Map