Assay Details
| Gene Symbol: prtfdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102892.1 | NP_001096362.1 | 4 - 5 | 384 | 99 | |
| XM_018094511.1 | XP_017950000.1 | 4 - 5 | 665 | 99 | ||
| XM_018094512.1 | XP_017950001.1 | 4 - 5 | 662 | 99 | ||
| GenBank mRNA | BC135868.1 | - | 4 - 5 | 384 | 99 | |
More Information
Set Membership: |
|

prtfdc1
Genomic Map