Assay Details
| Gene Symbol: trmt44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012955724.2 | XP_012811178.1 | 1 - 2 | 495 | 63 | |
| XM_012955725.2 | XP_012811179.1 | 1 - 2 | 525 | 63 | ||
| GenBank mRNA | BC135974.1 | - | 1 - 2 | 443 | 63 | |
| CT025237.2 | - | 1 - 2 | 411 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_012955725.2 transcripts.
Set Membership: |
|

trmt44
Genomic Map