Assay Details
| Gene Symbol: cbarp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102920.1 | NP_001096390.1 | 5 - 6 | 510 | 94 | |
| XM_012965443.2 | XP_012820897.1 | 4 - 5 | 591 | 94 | ||
| GenBank mRNA | BC135992.1 | - | 5 - 6 | 510 | 94 | |
More Information
Set Membership: |
|

cbarp
Genomic Map