Assay Details
| Gene Symbol: cbarp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102920.1 | NP_001096390.1 | 7 - 8 | 864 | 82 | |
| XM_012965443.2 | XP_012820897.1 | 6 - 7 | 945 | 82 | ||
| GenBank mRNA | BC135992.1 | - | 7 - 8 | 864 | 82 | |
More Information
Set Membership: |
|

cbarp
Genomic Map