Assay Details
| Gene Symbol: dnajb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102934.1 | NP_001096404.1 | 1 - 2 | 396 | 75 | |
| XM_018092893.1 | XP_017948382.1 | 1 - 2 | 791 | 75 | ||
| GenBank mRNA | BC136043.1 | - | 1 - 2 | 396 | 75 | |
More Information
Set Membership: |
|

dnajb4
Genomic Map