Assay Details
| Gene Symbol: mfhas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102937.1 | NP_001096407.1 | 1 - 2 | 2985 | 80 | |
| GenBank mRNA | BC136050.1 | - | 1 - 2 | 2985 | 80 | |
More Information
Set Membership: |
|

mfhas1
Genomic Map