Assay Details
| Gene Symbol: zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102939.1 | NP_001096409.1 | 7 - 8 | 2240 | 64 | |
| GenBank mRNA | BC136052.1 | - | 7 - 8 | 2240 | 64 | |
More Information
Set Membership: |
|

zbtb49
Genomic Map