Assay Details
| Gene Symbol: cacng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102948.1 | NP_001096418.1 | 1 - 2 | 330 | 95 | |
| GenBank mRNA | BC136081.1 | - | 1 - 2 | 330 | 95 | |
More Information
Set Membership: |
|

cacng2
Genomic Map